5-Chloro-2,3-diaminopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloropyridine-2,3-diyldiamine, 5-Chloropyridine-2,3-diamine, 2,3-Diamino-5-chloropyridine

35034-0050 35034-0250
TS35034-0050EA 30 USD
TS35034-0050 TS35034-0250
5-Chloro-2,3-diaminopyridine 97%
5-Chloro-2,3-diaminopyridine
Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Boiling Pt: 338.9 °C
Melting Pt: 170…175 °C
MDL Number: MFCD01230986
CAS Number: 25710-20-7

Order Now

Specification Test Results

Appearance White to grey to brown Powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR