5-Bromo-2,3-diaminopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine

35033-0050 35033-0250
TS35033-0050EA 18 USD
TS35033-0050 TS35033-0250
5-Bromo-2,3-diaminopyridine 97%
5-Bromo-2,3-diaminopyridine
Formula: C₅H₆BrN₃
MW: 188.03 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 160…165 °C
MDL Number: MFCD00460094
CAS Number: 38875-53-5
EINECS: 254-172-9

Order Now

Specification Test Results

Appearance Light yellow to light brown to purple Powder or needles
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR