(R)-(-)-Epichlorhydrin 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane
34901-0010
34901-0050
34901-0250
TS34901-0010EA
42
USD
TS34901-0010
TS34901-0050
TS34901-0250
(R)-(-)-Epichlorhydrin 99%
(R)-(-)-Epichlorhydrin
Formula:
ClCH₂CH(O)CH₂ MW: 92.52 g/mol Boiling Pt: 114 °C (760 mmHg) Melting Pt: –48 °C Density: 1.183 g/cm³ Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077759 CAS Number: 51594-55-9 EINECS: 424-280-2 UN: 2023 ADR: 6.1,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4360 to 1.4400 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific optical rotation | -34° to -39° (20°C, 589 nm) (c=1, MeOH) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...