4-Pyridone 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Hydroxypyridine, γ-Pyridone, 4-Pyridinol, Pyridin-4-ol, 1,4-Dihydro-4-Pyridinone

34883-0250 34883-1000 34883-5000
TS34883-0250EA 48 USD
TS34883-0250 TS34883-1000 TS34883-5000
4-Pyridone 95%
4-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 381
Melting Pt: 140…147 °C
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3

Order Now

Specification Test Results

Appearance White to yellow to beige to light brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 139 to 151 °C
HPLC ≥94 %
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR