8-hydroxyquinoline-2-carbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-Hydroxyquinoline-2-carbonitrile

34764-0010 34764-0050
TS34764-0010EA 52 USD
TS34764-0010 TS34764-0050
8-hydroxyquinoline-2-carbonitrile 98%
8-hydroxyquinoline-2-carbonitrile
Formula: C₁₀H₆N₂O
MW: 170.171 g/mol
Melting Pt: 131…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00216717
CAS Number: 6759-78-0

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 131 to 136 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR