8-Hydroxyquinoline-2-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-Hydroxyquinoline-2-carboxaldehyde

34763-0010 34763-0050
TS34763-0010EA 104 USD
TS34763-0010 TS34763-0050
8-Hydroxyquinoline-2-carbaldehyde 98%
8-Hydroxyquinoline-2-carbaldehyde
Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00168962
CAS Number: 14510-06-6

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 100 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR