8-hydroxyquinoline-2-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-Hydroxyquinoline-2-carboxylic acid

34762-0010 34762-0050
TS34762-0010EA 113 USD
TS34762-0010 TS34762-0050
8-hydroxyquinoline-2-carboxylic acid 98%
8-hydroxyquinoline-2-carboxylic acid
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 213…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00168972
CAS Number: 1571-30-8

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 219 to 223 °C
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR