2',7'-Dichlorofluorescein 3',6'-diacetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2',7'-Dichlorofluorescin diacetate, 2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester, 2',7'-Dichloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate, H₂DCFDA
34725-0010
34725-0050
TS34725-0010EA
115
USD
TS34725-0010
TS34725-0050
2',7'-Dichlorofluorescein 3',6'-diacetate 97%
2',7'-Dichlorofluorescein 3',6'-diacetate
Formula:
C₂₄H₁₄Cl₂O₇ Melting Pt: 239…246 °C Flash Pt: >109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037501 CAS Number: 2044-85-1 UN: 1759 ADR: 8,II |
Specification Test Results
Appearance | Colorless to off-white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 239 to 246 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...