Acriflavine hydrochloride

Supplier: Thermo Scientific Chemicals
Danger

34684-0100 34684-0250 34684-1000
TS34684-0100EA 45 USD
TS34684-0100 TS34684-0250 TS34684-1000
Acriflavine hydrochloride
Acriflavine hydrochloride
Formula: C₂₇H₂₈Cl₄N₆
MW: 578.36 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00069039
CAS Number: 8063-24-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Orange-red to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤7 % (105°C, 1 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR