1,2,3-Triazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3-1H-Triazole, 1H-1,2,3-Triazole

34548-0010 34548-0050 34548-0250
TS34548-0010EA 53 USD
TS34548-0010 TS34548-0050 TS34548-0250
1,2,3-Triazole 97%
1,2,3-Triazole
Formula: C₂H₃N₃
Boiling Pt: 203 °C (752 mmHg)
Melting Pt: 23…25 °C
Density: 1.192 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00014490
CAS Number: 288-36-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR