4-Amino-2-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloro-4-aminopyridine

34545-0050 34545-0250 34545-1000
TS34545-0050EA 61 USD
TS34545-0050 TS34545-0250 TS34545-1000
4-Amino-2-chloropyridine 97%
4-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00060089
CAS Number: 14432-12-3
EINECS: 238-403-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 89 to 94 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR