4-Aminopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

34507-0010 34507-0050
TS34507-0010EA 133 USD
TS34507-0010 TS34507-0050
4-Aminopyrimidine 98%
4-Aminopyrimidine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 151…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006112
CAS Number: 591-54-8

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 151 to 155 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR