2-Thiopheneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid

34471-0010 34471-0050 34471-0250
TS34471-0010EA 29 USD
TS34471-0010 TS34471-0050 TS34471-0250
2-Thiopheneboronic acid 97%
2-Thiopheneboronic acid
Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 120…122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151850
CAS Number: 6165-68-0

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR