Triallyl isocyanurate 98% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, Isocyanuric Acid Triallyl Ester, 1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione

34212-1000 34212-5000
TS34212-1000EA 61 USD
TS34212-1000 TS34212-5000
Triallyl isocyanurate 98% stabilised
Triallyl isocyanurate
Formula: C₁₂H₁₅N₃O₃
Boiling Pt: 149…152 °C (4 mmHg)
Melting Pt: 24.8 °C
Density: 1.159 g/cm³
Flash Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00006554
CAS Number: 1025-15-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid or solid
Infrared spectrum Conforms
Refractive index 1.5120 to 1.5140 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 100 to 1200 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR