1,2,2,6,6-Pentamethylpiperidin-4-amine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Amino-1,2,2,6,6-pentamethylpiperidine

33594-0010 33594-0050
TS33594-0010EA 71 USD
TS33594-0010 TS33594-0050
1,2,2,6,6-Pentamethylpiperidin-4-amine 99%
1,2,2,6,6-Pentamethylpiperidin-4-amine
Formula: C₁₀H₂₂N₂
Boiling Pt: 215…218 °C (760 mmHg)
Flash Pt: 72 °C
Storage Temperature: Ambient
CAS Number: 40327-96-6
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR