Piperonyl butoxide 90% tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole, 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole, NSC 8401

33416-1000 33416-5000
TS33416-1000EA 57 USD
TS33416-1000 TS33416-5000
Piperonyl butoxide 90% tech.
Piperonyl butoxide
Formula: C₁₉H₃₀O₅
MW: 338.44 g/mol
Boiling Pt: 180 °C (1 mmHg)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005842
CAS Number: 51-03-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4970 to 1.5070 (20°C, 589 nm)
GC ≥88 %
Specific gravity (20°C) 1.055 to 1.065

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR