(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone 98%

Supplier: Thermo Scientific Chemicals
Warning

33356-0010 33356-0050 33356-0250
TS33356-0010EA 28 USD
TS33356-0010 TS33356-0050 TS33356-0250
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone 98%
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
Formula: C₅H₉NO₂
Boiling Pt: 147…149 °C
Melting Pt: 77…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00077791
CAS Number: 66673-40-3

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 77 to 84 °C
HPLC ≥97.5 %
Specific optical rotation -27° to -35° (20 °C, 589 nm) (c=1, C2H5OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR