1-Methyl-1H-indole-3-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methylindole-3-carboxaldehyde

33161-0010 33161-0100
TS33161-0010EA 32 USD
TS33161-0010 TS33161-0100
1-Methyl-1H-indole-3-carbaldehyde 97%
1-Methyl-1H-indole-3-carbaldehyde
Formula: C₁₀H₉NO
MW: 159.188 g/mol
Melting Pt: 67…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00014570
CAS Number: 19012-03-4

Order Now

Specification Test Results

Appearance Light yellow to orange to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 67 to 72 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR