3',4'-(Methylenedioxy)acetophenone 98%

Supplier: Thermo Scientific Chemicals

33081-0050 33081-0250
TS33081-0050EA 38 USD
TS33081-0050 TS33081-0250
3',4'-(Methylenedioxy)acetophenone 98%
3',4'-(Methylenedioxy)acetophenone
Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 86…89 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005831
CAS Number: 3162-29-6
EINECS: 221-613-1

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 89 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR