3-(2-Furyl)propanoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Furan-2-yl)propanoic acid

33010-0010 33010-0100
TS33010-0010EA 46 USD
TS33010-0010 TS33010-0100
3-(2-Furyl)propanoic acid 98%
3-(2-Furyl)propanoic acid
Formula: C₇H₈O₃
MW: 140.138 g/mol
Melting Pt: 56…59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005346
CAS Number: 935-13-7

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 54 to 58 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR