1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one 98%

Supplier: Thermo Scientific Chemicals

Synonyms: ODQ

32880-0100 32880-0500
TS32880-0100EA 115 USD
TS32880-0100 TS32880-0500
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one 98%
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one
Formula: C₉H₅N₃O₂
Melting Pt: 160…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792620
CAS Number: 41443-28-1

Order Now

Specification Test Results

Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Melting point 160 to 170 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR