Wortmannin 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (1S,6bR,9aS,11R,11bR)-1-(Methoxymethyl)-9a,11b-dimethyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3H-furo[4,3,2-de]indeno[4,5-h]isochromen-11-yl acetate

32859-0010 32859-0050
TS32859-0010EA 109 USD
TS32859-0010 TS32859-0050
Wortmannin 98%
Wortmannin
Formula: C₂₃H₂₄O₈
MW: 428.44 g/mol
Melting Pt: 234…240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00133927
CAS Number: 19545-26-7
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Appearance White to light yellow Powder or solid or film
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR