2,2'-Bipyridyl-1,1'-dioxide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Dipyridyl N,N'-Dioxide, [2,2'-Bipyridine] 1,1'-dioxide, 2,2'-Bipyridine 1,1'-Dioxide

31961-0010 31961-0050
TS31961-0010EA 48 USD
TS31961-0010 TS31961-0050
2,2'-Bipyridyl-1,1'-dioxide 98%
2,2'-Bipyridyl-1,1'-dioxide
Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 296…298 °C
Storage Temperature: Ambient
MDL Number: MFCD00065169
CAS Number: 7275-43-6

Order Now

Specification Test Results

Appearance Off-white to cream Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR