2-Amino-4-methyl-3-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-3-nitro-2-pyridylamine, 4-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-4-picoline

31955-0250 319550050
TS31955-0250EA 120 USD
TS31955-0250 TS319550050
2-Amino-4-methyl-3-nitropyridine 98%
2-Amino-4-methyl-3-nitropyridine
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 140
Storage Temperature: Ambient
MDL Number: MFCD00006315
CAS Number: 6635-86-5
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 141 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR