2,2-Dimethylsuccinic anhydride 98%

Supplier: Thermo Scientific Chemicals
Warning

30876-0010 30876-0050
TS30876-0010EA 58 USD
TS30876-0010 TS30876-0050
2,2-Dimethylsuccinic anhydride 98%
2,2-Dimethylsuccinic anhydride
Formula: C₆H₈O₃
Boiling Pt: 219…220 °C
Melting Pt: 26…30 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00022608
CAS Number: 17347-61-4
EINECS: 241-371-0

Order Now

Specification Test Results

Appearance White Low melting solid
Infrared spectrum Conforms
Melting point 26 to 30 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR