6-Methylnicotinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid

30762-0010 30762-0050
TS30762-0010EA 33 USD
TS30762-0010 TS30762-0050
6-Methylnicotinic acid 99%
6-Methylnicotinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 209…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00006341
CAS Number: 3222-47-7

Order Now

Specification Test Results

Appearance White to light yellow to beige Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 209 to 213 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR