2,6-Difluoropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

30666-0050 30666-0250
TS30666-0050EA 41 USD
TS30666-0050 TS30666-0250
2,6-Difluoropyridine 99%
2,6-Difluoropyridine
Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 124.5 °C (743 mmHg)
Density: 1.26 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006227
CAS Number: 1513-65-1
EINECS: 216-152-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to pink to red-purple Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4390 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR