4-Methyl-1,3-dioxane 99%

Supplier: Thermo Scientific Chemicals
Danger

30500-0050 30500-0500
TS30500-0050EA 31 USD
TS30500-0050 TS30500-0500
4-Methyl-1,3-dioxane 99%
4-Methyl-1,3-dioxane
Formula: C₅H₁₀O₂
MW: 102.1317 g/mol
Melting Pt: 45…44 °C
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00006569
CAS Number: 1120-97-4
EINECS: 214-323-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4160 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR