1-Methylimidazolidin-2-one 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-2-imidazolidinone

30415-0020 30415-5000
TS30415-0020EA 291 USD
TS30415-0020 TS30415-5000
1-Methylimidazolidin-2-one 98+%
1-Methylimidazolidin-2-one
Formula: C₄H₈N₂O
MW: 100.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800601
CAS Number: 694-32-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline solid
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR