8-Methylquinoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
30362-0050
30362-0250
TS30362-0050EA
47
USD
TS30362-0050
TS30362-0250
8-Methylquinoline 97%
8-Methylquinoline
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 248 °C (1013 hPa) Melting Pt: –80 °C Density: 1.05 g/cm³ Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006810 CAS Number: 611-32-5 EINECS: 210-264-0 |
Specification Test Results
Appearance | Clear colorless to yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6130 to 1.6170 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...