(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone 97%

Supplier: Thermo Scientific Chemicals
Warning

30314-0010 30314-0050
TS30314-0010EA 23 USD
TS30314-0010 TS30314-0050
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone 97%
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 78…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00077792
CAS Number: 17342-08-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 78 to 86 °C
HPLC ≥96.0 %
Specific optical rotation +27.5° to +32.5° (20 °C, 589 nm) (c=2, C2H5OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR