3,4-(Methylenedioxy)phenylacetic acid 98.5+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1,3-Benzodioxol-5-yl)acetic acid

30262-0025 30262-0250
TS30262-0025EA 37 USD
TS30262-0025 TS30262-0250
3,4-(Methylenedioxy)phenylacetic acid 98.5+%
3,4-(Methylenedioxy)phenylacetic acid
Formula: C₉H₈O₄
MW: 180.16
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00014576
CAS Number: 2861-28-1
EINECS: 220-679-9

Order Now

Specification Test Results

Appearance Beige to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 132 °C
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.5 %
Titration with NaOH ≥98.50 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR