2-Thiophenecarbonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
29805-0100
29805-0500
29805-2500
TS29805-0100EA
65
USD
TS29805-0100
TS29805-0500
TS29805-2500
2-Thiophenecarbonitrile 97%
2-Thiophenecarbonitrile
Formula:
C₅H₃NS Boiling Pt: 192 °C (760 mmHg) Density: 1.17 g/cm³ Flash Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005416 CAS Number: 1003-31-2 EINECS: 213-706-0 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5620 to 1.5640 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...