2-(2-Bromoethyl)-1,3-dioxane 98%

Supplier: Thermo Scientific Chemicals
Warning

29793-0100 29793-0500
TS29793-0100EA 48 USD
TS29793-0100 TS29793-0500
2-(2-Bromoethyl)-1,3-dioxane 98%
2-(2-Bromoethyl)-1,3-dioxane
Formula: C₆H₁₁BrO₂
Boiling Pt: 67…70 °C (2.8 mmHg)
Density: 1.431 g/cm³
Flash Pt: 97 °C
Storage Temperature: Freezer
MDL Number: MFCD00006567
CAS Number: 33884-43-4

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4840 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR