N-Boc-3-(4-cyanophenyl)oxaziridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

29709-0020 29709-5000
TS29709-0020EA 1210 USD
TS29709-0020 TS29709-5000
N-Boc-3-(4-cyanophenyl)oxaziridine 98+%
N-Boc-3-(4-cyanophenyl)oxaziridine
Formula: C₁₃H₁₄N₂O₃
Melting Pt: 57…63 °C
Storage Temperature: Freezer
MDL Number: MFCD00800493
CAS Number: 150884-56-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 63 °C
Elemental Analysis ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR