(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%, >95% ee

Supplier: Thermo Scientific Chemicals
Danger

29704-0010 29704-0050
TS29704-0010EA 55 USD
TS29704-0010 TS29704-0050
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%, >95% ee
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula: C₆H₇NO
Melting Pt: 92…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00211274
CAS Number: 79200-56-9

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 92 to 100 °C
HPLC ≥97.5 %
UV at 400 nm A: ≤0.1
Enantiomeric excess ≥95 % (HPLC)
Specific optical rotation -550° to -570° (25 °C, 589 nm) (c=1, CH2Cl2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR