4-(4-Chlorophenyl)piperidin-4-ol 99+%, crystalline

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4-Chlorophenyl)-4-hydroxypiperidine

29684-0010 29684-0050 29684-0250
TS29684-0010EA 28 USD
TS29684-0010 TS29684-0050 TS29684-0250
4-(4-Chlorophenyl)piperidin-4-ol 99+%, crystalline
4-(4-Chlorophenyl)piperidin-4-ol
Formula: C₁₁H₁₄ClNO
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00006001
CAS Number: 39512-49-7
EINECS: 254-479-8

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 2 h, 760 mmHg)
Sulfated ash ≤0.2 %
TLC ≤0.5 % (Each individual impurity)
UV at 440 nm A: ≤0.05
Titration with HClO4 98.0 to 102.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR