3-Phenyl-1H-pyrazole 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole

29603-0025 29603-0100
TS29603-0025EA 53 USD
TS29603-0025 TS29603-0100
3-Phenyl-1H-pyrazole 99+%
3-Phenyl-1H-pyrazole
Formula: C₉H₈N₂
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00159654
CAS Number: 2458-26-6

Order Now

Specification Test Results

Appearance White to yellow to light orange Powder
Infrared spectrum Conforms
Melting point 74 to 80 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR