4,6-Dimethyl-2-pyrone 98%

Supplier: Thermo Scientific Chemicals

29535-0010 29535-0100
TS29535-0010EA 47 USD
TS29535-0010 TS29535-0100
4,6-Dimethyl-2-pyrone 98%
4,6-Dimethyl-2-pyrone
Formula: C₇H₈O₂
Melting Pt: 48…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075555
CAS Number: 675-09-2

Order Now

Specification Test Results

Appearance yellow Low melting crystalline mass (May darken on storage)
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR