6-Methylquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

29514-0050 29514-0250 29514-1000
TS29514-0050EA 23 USD
TS29514-0050 TS29514-0250 TS29514-1000
6-Methylquinoline 98%
6-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 256…260 °C (760 mmHg)
Density: 1.06 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006804
CAS Number: 91-62-3
EINECS: 202-084-6

Order Now

Specification Test Results

Appearance Light yellow to light orange Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR