3-Hydroxy-1,2-dimethyl-4(1H)-pyridone 99+%

Supplier: Thermo Scientific Chemicals
Warning

27874-0050 27874-0100 27874-0500
TS27874-0050EA 72 USD
TS27874-0050 TS27874-0100 TS27874-0500
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone 99+%
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone
Formula: C₇H₉NO₂
MW: 139.15 g/mol
Melting Pt: 271…273 °C
Storage Temperature: Ambient
MDL Number: MFCD00134497
CAS Number: 30652-11-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or needles
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Heavy metals ≤20 ppm
Sulfated ash ≤0.1 %
TLC ≥99.0 %
pH 6 to 8 (1 % solution)
Titration with HClO4 ≥99.0 % (On dry substance)
Chloride (Cl) ≤100 ppm
Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR