2-Amino-1-methylbenzimidazole 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1H-benzo[d]imidazol-2-amine

27630-0025 27630-0100
TS27630-0025EA 128 USD
TS27630-0025 TS27630-0100
2-Amino-1-methylbenzimidazole 99+%
2-Amino-1-methylbenzimidazole
Formula: C₈H₉N₃
MW: 147.181 g/mol
Melting Pt: 203…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00142855
CAS Number: 1622-57-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 203 to 206 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR