4,4'-Bipyridyl, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
27620-0050
27620-0250
27620-1000
TS27620-0050EA
44
USD
TS27620-0050
TS27620-0250
TS27620-1000
4,4'-Bipyridyl, anhydrous 98%
4,4'-Bipyridyl
Formula:
C₁₀H₈N₂ Boiling Pt: 305 °C (760 mmHg) Melting Pt: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006416 CAS Number: 553-26-4 EINECS: 209-036-3 UN: 2811 ADR: 6.1,III Merck Index: 13,03383 |
Specification Test Results
Appearance | Off-white to beige Powder |
Infrared spectrum | Conforms |
Melting point | 110 to 114 °C |
Water | ≤2.0 % (K.F.) |
Sulfated ash | ≤0.1 % |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...