2-Mercapto-5-methoxybenzimidazole 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methoxy-2-benzimidazolethiol, 1,3-Dihydro-5-methoxy-2H-benzimidazole-2-thione, 5-Methoxy-1H-benzo[d]imidazole-2-thiol, 5-Methoxybenzimidazole-2-thiol, 2-Mercapto-5-methoxy-1H-benzimidazole

27537-0050 27537-0250
TS27537-0050EA 37 USD
TS27537-0050 TS27537-0250
2-Mercapto-5-methoxybenzimidazole 99+%
2-Mercapto-5-methoxybenzimidazole
Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 255…262 °C
MDL Number: MFCD00134581
CAS Number: 37052-78-1
EINECS: 253-326-2

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 255 to 260 °C
HPLC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR