ɑ-Hydroxy-γ-butyrolactone 99%

Supplier: Thermo Scientific Chemicals
Warning

27507-0050 27507-0250
TS27507-0050EA 119 USD
TS27507-0050 TS27507-0250
ɑ-Hydroxy-γ-butyrolactone 99%
alpha-Hydroxy-gamma-butyrolactone
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 85…90 °C (.8mbar)
Density: 1.313 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00134268
CAS Number: 19444-84-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR