3-Amino-5-hydroxy-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one

27172-0050 27172-0250 27172-1000
TS27172-0050EA 36 USD
TS27172-0050 TS27172-0250 TS27172-1000
3-Amino-5-hydroxy-1H-pyrazole 98%
3-Amino-5-hydroxy-1H-pyrazole
Formula: C₃H₅N₃O
MW: 99.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022384
CAS Number: 6126-22-3

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR