5-Hydroxysophthalic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxyisophthalic acid

27139-0250 27139-1000 27139-5000
TS27139-0250EA 43 USD
TS27139-0250 TS27139-1000 TS27139-5000
5-Hydroxysophthalic acid 99%
5-Hydroxysophthalic acid
Formula: C₈H₆O₅
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: 297…303 °C
MDL Number: MFCD00002515
CAS Number: 618-83-7
EINECS: 210-565-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 297 to 303 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR