2-Chloro-6-methylnicotinic acid 98+%

Supplier: Thermo Scientific Chemicals

27081-0050 27081-0250 27081-1000
TS27081-0050EA 42 USD
TS27081-0050 TS27081-0250 TS27081-1000
2-Chloro-6-methylnicotinic acid 98+%
2-Chloro-6-methylnicotinic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00134165
CAS Number: 30529-70-5
EINECS: 250-229-7

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 157 to 161 °C
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR