2,3-Lutidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dimethylpyridine

26876-0250 26876-1000
TS26876-0250EA 27 USD
TS26876-0250 TS26876-1000 TS26876-5000
2,3-Lutidine 98+%
2,3-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 161 °C (760 mmHg)
Melting Pt: –17 °C
Density: 0.95 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009605
CAS Number: 583-61-9
EINECS: 209-514-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5062 to 1.5082 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR