1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6,7-Dimethoxy-1-methyl-3,4-dihydroisoquinoline

26597-0010 26597-0050
TS26597-0010EA 144 USD
TS26597-0010 TS26597-0050
1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline 99+%
1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline
Formula: C₁₂H₁₅NO₂
MW: 205.257 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00040276
CAS Number: 4721-98-6

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR